Assay Details
| Gene Symbol: CLIC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039166.1 | XP_017894655.1 | 3 - 4 | 646 | 71 | |
| GenBank mRNA | JO376424.2 | - | 4 - 5 | 4151 | 71 | |
More Information
Set Membership: |
|

CLIC5
Genomic Map