Assay Details
| Gene Symbol: CNNM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041666.1 | XP_017897155.1 | 8 - 9 | 3581 | 56 | |
| XM_018041667.1 | XP_017897156.1 | 8 - 9 | 3581 | 56 | ||
| XM_018041668.1 | XP_017897157.1 | 7 - 8 | 3518 | 56 | ||
| XM_018041669.1 | XP_017897158.1 | 7 - 8 | 3518 | 56 | ||
More Information
Set Membership: |
|

CNNM1
Genomic Map