Assay Details
| Gene Symbol: DUSP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044792.1 | XP_017900281.1 | 3 - 4 | 1032 | 79 | |
| XM_018044793.1 | XP_017900282.1 | 3 - 4 | 1050 | 79 | ||
More Information
Set Membership: |
|

DUSP9
Genomic Map