Assay Details
| Gene Symbol: CCNQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044785.1 | XP_017900274.1 | 2 - 3 | 453 | 73 | |
| XM_018044786.1 | XP_017900275.1 | 2 - 3 | 450 | 73 | ||
| XM_018044787.1 | XP_017900276.1 | 2 - 3 | 450 | 73 | ||
| XM_018044788.1 | XP_017900277.1 | 3 - 4 | 448 | 73 | ||
| XM_018044789.1 | XP_017900278.1 | 2 - 3 | 306 | 73 | ||
| XM_018044790.1 | XP_017900279.1 | 2 - 3 | 289 | 73 | ||
| GenBank mRNA | JT650956.1 | - | 2 - 3 | 208 | 73 | |
More Information
Set Membership: |
|

CCNQ
Genomic Map