Assay Details
| Gene Symbol: PMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675860.2 | XP_005675917.1 | 6 - 7 | 827 | 92 | |
| XM_013967766.2 | XP_013823220.1 | 5 - 6 | 644 | 92 | ||
More Information
Set Membership: |
|

PMS1
Genomic Map