Assay Details
| Gene Symbol: RPRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063163.1 | XP_017918652.1 | 1 | 398 | 92 | |
| GenBank mRNA | JT748328.1 | - | 2 | 317 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RPRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063163.1 | XP_017918652.1 | 1 | 398 | 92 | |
| GenBank mRNA | JT748328.1 | - | 2 | 317 | 92 | |
Set Membership: |
|