Assay Details
| Gene Symbol: ZFAND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676560.2 | XP_005676617.1 | 6 - 7 | 615 | 55 | |
| XM_005676561.3 | XP_005676618.1 | 6 - 7 | 632 | 55 | ||
| XM_005676562.2 | XP_005676619.1 | 6 - 7 | 615 | 55 | ||
| XM_005676563.2 | XP_005676620.1 | 6 - 7 | 603 | 55 | ||
| XM_013972816.2 | XP_013828270.1 | 6 - 7 | 620 | 55 | ||
More Information
Set Membership: |
|

ZFAND2B
Genomic Map