Assay Details
| Gene Symbol: C3H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677458.3 | XP_005677515.1 | 2 - 3 | 398 | 61 | |
| GenBank mRNA | JO568686.2 | - | 2 - 3 | 525 | 61 | |
More Information
Set Membership: |
|

C3H1orf189
Genomic Map