Assay Details
| Gene Symbol: LMBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963627.2 | XP_013819081.1 | 3 - 4 | 179 | 63 | |
| XM_018046643.1 | XP_017902132.1 | 3 - 4 | 410 | 63 | ||
| XM_018046644.1 | XP_017902133.1 | 3 - 4 | 410 | 63 | ||
More Information
Set Membership: |
|

LMBR1
Genomic Map