Assay Details
| Gene Symbol: DNAJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682254.3 | XP_005682311.1 | 2 - 3 | 1001 | 82 | |
| GenBank mRNA | KA334866.1 | - | 1 - 2 | 1411 | 82 | |
More Information
Set Membership: |
|

DNAJB1
Genomic Map