Assay Details
| Gene Symbol: DNASE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682283.3 | XP_005682340.1 | 2 - 3 | 542 | 62 | |
| GenBank mRNA | JO546507.2 | - | 1 - 2 | 49 | 62 | |
More Information
Set Membership: |
|

DNASE2
Genomic Map