Assay Details
| Gene Symbol: LOC102174838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683355.2 | XP_005683412.1 | 4 - 5 | 421 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102174838| Gene Symbol: LOC102174838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683355.2 | XP_005683412.1 | 4 - 5 | 421 | 73 | |
Set Membership: |
|