Assay Details
| Gene Symbol: DMAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683677.3 | XP_005683734.2 | 1 - 2 | 333 | 57 | |
| GenBank mRNA | JO622344.2 | - | 1 - 2 | 295 | 57 | |
More Information
Set Membership: |
|

DMAC1
Genomic Map