Assay Details
| Gene Symbol: PSMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685017.3 | XP_005685074.1 | 2 - 3 | 277 | 66 | |
| GenBank mRNA | JO515129.2 | - | 2 - 3 | 293 | 66 | |
| JO632445.2 | - | 2 - 3 | 278 | 66 | ||
More Information
Set Membership: |
|

PSMB1
Genomic Map