Assay Details
| Gene Symbol: KLHL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687646.3 | XP_005687703.1 | 9 - 10 | 2828 | 69 | |
| XM_005687647.3 | XP_005687704.2 | 8 - 9 | 1839 | 69 | ||
More Information
Set Membership: |
|

KLHL1
Genomic Map