Assay Details
| Gene Symbol: POP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691570.3 | XP_005691627.2 | 2 - 3 | 252 | 56 | |
| GenBank mRNA | KA327444.1 | - | 2 - 3 | 208 | 56 | |
More Information
Set Membership: |
|

POP5
Genomic Map