Assay Details
| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064220.1 | XP_017919709.1 | 2 - 3 | 407 | 95 | |
| GenBank mRNA | JO437104.2 | - | 2 - 3 | 794 | 95 | |
| JO633299.2 | - | 2 - 3 | 618 | 95 | ||
| JO633300.2 | - | 2 - 3 | 618 | 95 | ||
More Information
Set Membership: |
|

RASL10B
Genomic Map