Assay Details
| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064220.1 | XP_017919709.1 | 3 - 4 | 540 | 73 | |
| GenBank mRNA | JO437104.2 | - | 1 - 2 | 661 | 73 | |
| JO633299.2 | - | 3 - 4 | 751 | 73 | ||
| JO633300.2 | - | 3 - 4 | 751 | 73 | ||
More Information
Set Membership: |
|

RASL10B
Genomic Map