Assay Details
| Gene Symbol: GABARAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693485.3 | XP_005693542.1 | 1 - 2 | 252 | 84 | |
| GenBank mRNA | JO581681.1 | - | 1 - 2 | 814 | 84 | |
More Information
Set Membership: |
|

GABARAP
Genomic Map