Assay Details
| Gene Symbol: G6PC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693877.3 | XP_005693934.1 | 4 - 5 | 735 | 81 | |
| XM_005693878.3 | XP_005693935.2 | 4 - 5 | 563 | 81 | ||
| GenBank mRNA | JO432881.2 | - | 4 - 5 | 775 | 81 | |
More Information
Set Membership: |
|

G6PC
Genomic Map