Assay Details
| Gene Symbol: MESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695171.3 | XP_005695228.3 | 2 - 3 | 519 | 67 | |
| GenBank mRNA | KA344909.1 | - | 1 - 2 | 1273 | 67 | |
More Information
Set Membership: |
|

MESD
Genomic Map