Assay Details
| Gene Symbol: LOC100861326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696862.3 | XP_005696919.2 | 2 - 3 | 1463 | 95 | |
| XM_005696863.3 | XP_005696920.2 | 2 - 3 | 1687 | 95 | ||
| GenBank mRNA | JO379140.2 | - | 2 - 3 | 1233 | 95 | |
| Gene Symbol: LOC106503512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JT685336.1 | - | 3 - 4 | 380 | 95 | |
More Information
Set Membership: |
|

LOC100861326
Genomic Map