Assay Details
| Gene Symbol: PRR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048890.1 | XP_017904379.1 | 4 - 5 | 456 | 58 | |
| XM_018048891.1 | XP_017904380.1 | 5 - 6 | 347 | 58 | ||
| GenBank mRNA | JO634055.2 | - | 4 - 5 | 1339 | 58 | |
More Information
Set Membership: |
|

PRR5
Genomic Map