Assay Details
| Gene Symbol: KANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050950.1 | XP_017906439.1 | 5 - 6 | 2154 | 72 | |
| XM_018050952.1 | XP_017906441.1 | 3 - 4 | 1857 | 72 | ||
| XM_018050953.1 | XP_017906442.1 | 5 - 6 | 3480 | 72 | ||
| GenBank mRNA | JO645107.2 | - | 5 - 6 | 1088 | 72 | |
More Information
Set Membership: |
|

KANK3
Genomic Map