Assay Details
| Gene Symbol: RNF44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050643.1 | XP_017906132.1 | 7 - 8 | 4084 | 57 | |
| XM_018050644.1 | XP_017906133.1 | 7 - 8 | 1209 | 57 | ||
| XM_018050645.1 | XP_017906134.1 | 7 - 8 | 3288 | 57 | ||
| XM_018050646.1 | XP_017906135.1 | 7 - 8 | 1065 | 57 | ||
| XM_018050647.1 | XP_017906136.1 | 7 - 8 | 1368 | 57 | ||
| XM_018050649.1 | XP_017906138.1 | 7 - 8 | 1873 | 57 | ||
| XM_018050650.1 | XP_017906139.1 | 6 - 7 | 930 | 57 | ||
| XM_018050651.1 | XP_017906140.1 | 7 - 8 | 1436 | 57 | ||
| XM_018050652.1 | XP_017906141.1 | 7 - 8 | 4016 | 57 | ||
| XM_018050653.1 | XP_017906142.1 | 7 - 8 | 1140 | 57 | ||
| GenBank mRNA | JO623862.2 | - | 7 - 8 | 1222 | 57 | |
| JT714302.1 | - | 2660 | 57 | |||
More Information
Set Membership: |
|

RNF44
Genomic Map