Assay Details
| Gene Symbol: HNRNPAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050576.1 | XP_017906065.1 | 3 - 4 | 1125 | 73 | |
| XM_018050577.1 | XP_017906066.1 | 3 - 4 | 958 | 73 | ||
| XM_018050578.1 | XP_017906067.1 | 3 - 4 | 1125 | 73 | ||
More Information
Set Membership: |
|

HNRNPAB
Genomic Map