Assay Details
| Gene Symbol: ADAMTSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050419.1 | XP_017905908.1 | 5 - 6 | 768 | 69 | |
| XM_018050420.1 | XP_017905909.1 | 5 - 6 | 767 | 69 | ||
| XM_018050421.1 | XP_017905910.1 | 5 - 6 | 761 | 69 | ||
| XM_018050422.1 | XP_017905911.1 | 5 - 6 | 637 | 69 | ||
| XM_018050423.1 | XP_017905912.1 | 5 - 6 | 765 | 69 | ||
| XM_018050424.1 | XP_017905913.1 | 5 - 6 | 761 | 69 | ||
| XM_018050425.1 | XP_017905914.1 | 5 - 6 | 772 | 69 | ||
| XM_018050426.1 | XP_017905915.1 | 5 - 6 | 635 | 69 | ||
| XM_018050427.1 | XP_017905916.1 | 5 - 6 | 638 | 69 | ||
| XM_018050428.1 | XP_017905917.1 | 3 - 4 | 268 | 69 | ||
| XM_018050429.1 | XP_017905918.1 | 4 - 5 | 323 | 69 | ||
| GenBank mRNA | JO393685.2 | - | 4 - 5 | 354 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018050427.1, XM_018050428.1, XM_018050429.1 transcripts.
Set Membership: |
|

ADAMTSL5
Genomic Map