Assay Details
| Gene Symbol: LOC100861136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064564.1 | XP_017920053.1 | 2 - 3 | 517 | 80 | |
| GenBank mRNA | JI861856.1 | - | 2 - 3 | 80 | ||
More Information
Set Membership: |
|

LOC100861136
Genomic Map