Assay Details
| Gene Symbol: HOXB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064930.1 | XP_017920419.1 | 1 - 2 | 1001 | 92 | |
| GenBank mRNA | JO454965.2 | - | 1 - 2 | 174 | 92 | |
More Information
Set Membership: |
|

HOXB8
Genomic Map