Assay Details
| Gene Symbol: CMTM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038286.1 | XP_017893775.1 | 2 - 3 | 387 | 60 | |
| GenBank mRNA | JO340842.2 | - | 2 - 3 | 349 | 60 | |
| JO426996.1 | - | 2 - 3 | 2515 | 60 | ||
| JO427001.1 | - | 2 - 3 | 2514 | 60 | ||
More Information
Set Membership: |
|

CMTM6
Genomic Map