Assay Details
| Gene Symbol: CNPY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038610.1 | XP_017894099.1 | 3 - 4 | 507 | 58 | |
| GenBank mRNA | JO501667.2 | - | 3 - 4 | 461 | 58 | |
More Information
Set Membership: |
|

CNPY3
Genomic Map