Assay Details
| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056663.1 | XP_017912152.1 | 3 - 4 | 354 | 58 | |
| XM_018056672.1 | XP_017912161.1 | 3 - 4 | 360 | 58 | ||
| GenBank mRNA | JO433308.2 | - | 3 - 4 | 1279 | 58 | |
| KA315773.1 | - | 3 - 4 | 405 | 58 | ||
More Information
Set Membership: |
|

TMEM222
Genomic Map