Assay Details
| Gene Symbol: BTN2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038551.1 | XP_017894040.1 | 3 - 4 | 808 | 64 | |
| GenBank mRNA | JO580544.1 | - | 785 | 64 | ||
| JO580545.1 | - | 3 - 4 | 785 | 64 | ||
More Information
Set Membership: |
|

BTN2A2
Genomic Map