Assay Details
| Gene Symbol: FGF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041114.1 | XP_017896603.1 | 5 - 6 | 641 | 80 | |
| XM_018041115.1 | XP_017896604.1 | 5 - 6 | 476 | 80 | ||
| XM_018041116.1 | XP_017896605.1 | 4 - 5 | 422 | 80 | ||
| XM_018041117.1 | XP_017896606.1 | 4 - 5 | 389 | 80 | ||
More Information
Set Membership: |
|

FGF8
Genomic Map