Assay Details
| Gene Symbol: MFHAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041952.1 | XP_017897441.1 | 1 - 2 | 3106 | 58 | |
| XM_018041953.1 | XP_017897442.1 | 1 - 2 | 3106 | 58 | ||
More Information
Set Membership: |
|

MFHAS1
Genomic Map