Assay Details
Gene Symbol: LOC108633215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043671.1 | XP_017899160.1 | 9 - 10 | 844 | 73 |
Gene Symbol: LOC102180115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO420746.2 | - | 1375 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
