Assay Details
Gene Symbol: LOC102180115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043681.1 | XP_017899170.1 | 750 | 68 | ||
GenBank mRNA | JO420746.2 | - | 609 | 68 |
Gene Symbol: LOC108633215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043671.1 | XP_017899160.1 | 2 - 3 | 78 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
