Assay Details
| Gene Symbol: KCNN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046203.1 | XP_017901692.1 | 7 - 8 | 2233 | 70 | |
| XM_018046204.1 | XP_017901693.1 | 7 - 8 | 2233 | 70 | ||
More Information
Set Membership: |
|

KCNN3
Genomic Map