Assay Details
| Gene Symbol: ACTRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675341.3 | XP_005675398.1 | 3 - 4 | 481 | 63 | |
| GenBank mRNA | KA318250.1 | - | 1 - 2 | 167 | 63 | |
More Information
Set Membership: |
|

ACTRT3
Genomic Map