Assay Details
| Gene Symbol: ORMDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065580.1 | XP_017921069.1 | 3 - 4 | 465 | 102 | |
| XM_018065586.1 | XP_017921075.1 | 3 - 4 | 440 | 102 | ||
| GenBank mRNA | JI861871.1 | - | 2 - 3 | 330 | 102 | |
| JO405987.2 | - | 3 - 4 | 374 | 102 | ||
More Information
Set Membership: |
|

ORMDL1
Genomic Map