Assay Details
| Gene Symbol: HNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676183.3 | XP_005676240.1 | 4 - 5 | 637 | 73 | |
| GenBank mRNA | JO507223.2 | - | 2 - 3 | 454 | 73 | |
More Information
Set Membership: |
|

HNMT
Genomic Map