Assay Details
| Gene Symbol: CDCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678679.3 | XP_005678736.1 | 3 - 4 | 347 | 59 | |
| GenBank mRNA | JO598349.2 | - | 8 - 9 | 1048 | 59 | |
| JT648404.1 | - | 11 - 12 | 810 | 59 | ||
| KA350976.1 | - | 2 - 3 | 254 | 59 | ||
More Information
Set Membership: |
|

CDCA8
Genomic Map