Assay Details
| Gene Symbol: LOC102175959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052330.1 | XP_017907819.1 | 6 - 7 | 1067 | 73 | |
| GenBank mRNA | JO335891.2 | - | 6 - 7 | 777 | 73 | |
More Information
Set Membership: |
|

LOC102175959
Genomic Map