Assay Details
| Gene Symbol: LOC102177447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686981.2 | XP_005687038.2 | 7 - 8 | 1204 | 86 | |
| XM_013967874.2 | XP_013823328.2 | 7 - 8 | 1174 | 86 | ||
| XM_013967875.2 | XP_013823329.2 | 7 - 8 | 1120 | 86 | ||
More Information
Set Membership: |
|

LOC102177447
Genomic Map