Assay Details
| Gene Symbol: SPAG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057784.1 | XP_017913273.1 | 8 - 9 | 907 | 65 | |
| XM_018057785.1 | XP_017913274.1 | 8 - 9 | 923 | 65 | ||
| XM_018057786.1 | XP_017913275.1 | 8 - 9 | 894 | 65 | ||
| GenBank mRNA | KA331748.1 | - | 1 - 2 | 40 | 65 | |
More Information
Set Membership: |
|

SPAG4
Genomic Map