Assay Details
| Gene Symbol: TMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690286.3 | XP_005690343.1 | 5 - 6 | 687 | 67 | |
| XM_005690287.3 | XP_005690344.1 | 5 - 6 | 632 | 67 | ||
More Information
Set Membership: |
|

TMX2
Genomic Map