Assay Details
| Gene Symbol: ADORA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690366.3 | XP_005690423.1 | 2 - 3 | 575 | 75 | |
| GenBank mRNA | JO645696.2 | - | 1 - 2 | 2788 | 75 | |
| JO645697.2 | - | 1 - 2 | 2788 | 75 | ||
More Information
Set Membership: |
|

ADORA1
Genomic Map