Assay Details
| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693150.3 | XP_005693207.2 | 2 - 3 | 222 | 63 | |
| GenBank mRNA | JO629855.2 | - | 2 - 3 | 213 | 63 | |
| JO629856.2 | - | 2 - 3 | 213 | 63 | ||
More Information
Set Membership: |
|

ZNHIT3
Genomic Map