Assay Details
| Gene Symbol: YES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697065.3 | XP_005697122.1 | 5 - 6 | 805 | 77 | |
| XM_018039696.1 | XP_017895185.1 | 5 - 6 | 784 | 77 | ||
| XM_018039697.1 | XP_017895186.1 | 5 - 6 | 698 | 77 | ||
| XM_018039698.1 | XP_017895187.1 | 5 - 6 | 697 | 77 | ||
More Information
Set Membership: |
|

YES1
Genomic Map