Assay Details
| Gene Symbol: POLL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041143.1 | XP_017896632.1 | 6 - 7 | 1157 | 66 | |
| XM_018041144.1 | XP_017896633.1 | 6 - 7 | 1412 | 66 | ||
| XM_018041145.1 | XP_017896634.1 | 5 - 6 | 857 | 66 | ||
More Information
Set Membership: |
|

POLL
Genomic Map